Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529100 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529100 Supplier Invitrogen™ Supplier No. PA529100
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, NIH-3T3. Predicted reactivity: Mouse (95%), Rat (95%), Rabbit (92%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the cytosolic phospholipase A2 group IV family. The enzyme catalyzes the hydrolysis of membrane phospholipids to release arachidonic acid which is subsequently metabolized into eicosanoids. Eicosanoids, including prostaglandins and leukotrienes, are lipid-based cellular hormones that regulate hemodynamics, inflammatory responses, and other intracellular pathways. The hydrolysis reaction also produces lysophospholipids that are converted into platelet-activating factor. The enzyme is activated by increased intracellular Ca(2+) levels and phosphorylation, resulting in its translocation from the cytosol and nucleus to perinuclear membrane vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G4A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene PLA2G4A
Gene Accession No. P47712, P47713
Gene Alias calcium-dependent phospholipid-binding protein; Cpla2; cPLA2alpha; cPLA2-alpha; cytosolic group IVA; Cytosolic phospholipase A2; group IVA; lysophospholipase; MGC126350; Phosphatidylcholine 2-acylhydrolase; Phospholipase A2; phospholipase A2 cytosolic; phospholipase A2 group IVA; phospholipase A2, group IVA (cytosolic, calcium-dependent); Pla2c; Pla2g4; PLA2G4A
Gene Symbols PLA2G4A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human PLA2G4A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18783, 5321
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.