Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G5 Monoclonal Antibody (3G1)
GREENER_CHOICE

Catalog No. 01674565 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-565 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-565 Supplier Invitrogen™ Supplier No. MA1830
  • $602.55
    $481.65 / Each of 1

    Save $120.90

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-830 contains 100 μg of in vitro produced, protein A purified antibody (1 mg/mL) in PBS containing 1 mg/mL BSA and 0.05% sodium azide. MA1-830 detects Group V Secretory Phospholipase A2 from human and mouse samples. MA1-830 has been successfully used in Western blot procedures. By Western blot, MA1-830 detects a ~15 kDa band representing Group V Secretory Phospholipase A2 from mouse macrophages. The MA1-830 immunogen is full length W79A human group V phospholipase A2.

This gene is a member of the secretory phospholipase A2 family. It is located in a tightly-linked cluster of secretory phospholipase A2 genes on chromosome 1. The encoded enzyme catalyzes the hydrolysis of membrane phospholipids to generate lysophospholipids and free fatty acids including arachidonic acid. It preferentially hydrolyzes linoleoyl-containing phosphatidylcholine substrates. Secretion of this enzyme is thought to induce inflammatory responses in neighboring cells. Alternatively spliced transcript variants have been found, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G5
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3G1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene PLA2G5
Gene Accession No. P39877, P97391
Gene Alias Ca2+-dependent phospholipase A2; calcium-dependent phospholipase A2; DKFZp686C2294; FRFB; group V phospholipase A2; GV-PLA2; hVPLA(2); MGC46205; Phosphatidylcholine 2-acylhydrolase 5; phospholipase A2 group V; phospholipase A2 group V; calcium-dependent phospholipase A2; phospholipase A2, group 5; phospholipase A2, group V; PLA2; PLA2-10; Pla2g5; sPLA2
Gene Symbols PLA2G5
Host Species Mouse
Immunogen Full length W79A human group V phospholipase A2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18784, 5322
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.