Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584150 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584150 Supplier Invitrogen™ Supplier No. PA584150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: GLLDLKSMIE KVTGKNALTN YGFYGCYC Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 86%.

This gene is a member of the secretory phospholipase A2 family. It is located in a tightly-linked cluster of secretory phospholipase A2 genes on chromosome 1. The encoded enzyme catalyzes the hydrolysis of membrane phospholipids to generate lysophospholipids and free fatty acids including arachidonic acid. It preferentially hydrolyzes linoleoyl-containing phosphatidylcholine substrates. Secretion of this enzyme is thought to induce inflammatory responses in neighboring cells. Alternatively spliced transcript variants have been found, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLA2G5
Gene Accession No. P39877
Gene Alias Ca2+-dependent phospholipase A2; calcium-dependent phospholipase A2; DKFZp686C2294; FRFB; group V phospholipase A2; GV-PLA2; hVPLA(2); MGC46205; Phosphatidylcholine 2-acylhydrolase 5; phospholipase A2 group V; phospholipase A2 group V; calcium-dependent phospholipase A2; phospholipase A2, group 5; phospholipase A2, group V; PLA2; PLA2-10; Pla2g5; sPLA2
Gene Symbols PLA2G5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLA2G5. Recombinant protein control fragment (Product #RP-99794).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5322
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.