Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2R1 Monoclonal Antibody (CL0485)
GREENER_CHOICE

Catalog No. PIMA524608 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524608 Supplier Invitrogen™ Supplier No. MA524608
  • $630.00
    $498.65 / Each of 1

    Save $131.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Immunogen sequence: EEKTWHEALR SCQADNSALI DITSLAEVEF LVTLLGDENA SETWIGLSSN KIPVSFEWSN DSSVIFTNWH TLEPHIFPNR SQLCVSAEQS EGHWKVKNCE ERLFYICKKA GHVLSDAESG CQEGWERHGG FCYKID Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%. Binds to an epitope located within the peptide sequence VKNCEERLFYICKKA as determined by overlapping synthetic peptides.

Receptor for secretory phospholipase A2 (sPLA2). Acts as a receptor for phosholipase sPLA2-IB/PLA2G1B but not sPLA2-IIA/PLA2G2A. Also able to bind to snake PA2-like toxins. Although its precise function remains unclear, binding of sPLA2 to its receptor participates in both positive and negative regulation of sPLA2 functions as well as clearance of sPLA2. Binding of sPLA2-IB/PLA2G1B induces various effects depending on the cell type, such as activation of the mitogen-activated protein kinase (MAPK) cascade to induce cell proliferation, the production of lipid mediators, selective release of arachidonic acid in bone marrow-derived mast cells. In neutrophils, binding of sPLA2-IB/PLA2G1B can activate p38 MAPK to stimulate elastase release and cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2R1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL0485
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pla2r1
Gene Accession No. Q13018
Gene Alias 180 kDa secretory phospholipase A2 receptor; CLEC13C; C-type lectin domain family 13 member C; mannose receptor, C type 2; Mrc2; M-type receptor; phospholipase A2 receptor 1; phospholipase A2 receptor 1, 180kD; phospholipase A2 receptor 1, 180kDa; phospholipase A2, group IB, pancreas, receptor; Pla2g1br; PLA2G1R; PLA2-I receptor; PLA2IR; Pla2r; Pla2-r; PLA2R1; Secretory phospholipase A2 receptor; Soluble PLA2-R; Soluble secretory phospholipase A2 receptor
Gene Symbols Pla2r1
Host Species Mouse
Immunogen Recombinant protein fragment of Human PLA2R1 (Product #RP-90366)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22925
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.