Learn More
Eisco™ Plain Spherical Mirror
SCI_ED
Catalog No. S24358 Shop All Eisco Products
Change view
Click to view available options
Diameter (Metric) Lens:
75 mm
100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Lens
S24358 100 mm
S24357 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S24358 Supplier Eisco™ Supplier No. PH0527D
Add to Cart
Add to Cart
This item is not returnable. View return policy
Plain Spherical Mirror with optically true glass.
  • With scratch-resistant copper-back silvering.
Specifications
Product Type Mirror
Lens Material Optically True Glass
Includes Scratch resistant copper back silvering
Diameter (English) Lens 3.94 in.
Diameter (Metric) Lens 100 mm