Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Planck Constant Determination Box
SCI_ED

Catalog No. S24913 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W):
4.75 x 8 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S24913 4.75 x 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24913 Supplier Eisco™ Supplier No. PH1365
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For determining the approximate value of Plancks constant using elbow voltage.

  • Utilizes elbow voltage in characteristic curves of LEDs
  • Seven LEDs covering from deep blue at 470nm to 940nm mounted on box
  • External power supply of 9V required or 9V battery with connecting cords

Specifications

Product Type Planck's Constant Determination Box
For Use With (Application) Determining the approximate value of Planck's constant
Includes Plastic viewing tube
Required Accessories Power Supply (9V DC), Voltmeter (0-9 Volts), Ammeter (20mA) and 6 Banana Plug Connecting Leads
Dimensions (L x W) 4.75 x 8 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.