Learn More
LAMDA BIOTECH INC PLANT-DIRECT™PCR Kit
Supplier:  LAMDA BIOTECH INC D302100
Encompass_Preferred
Plant-DirectTM PCR Kit contains all the reagents needed for quick preparation of plant genomic DNA and PCR amplification with the prepared DNA samples. Any plant tissues can be used such as leaf, stem, root or cultured plant tissues. This kit can be applied to any plant species as long as correct primers are used. The PCR products can be directly loaded onto agarose gel without addition of loading dye for visualization of the experimental results.
Catalog No. 50-204-5783
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.