Learn More
Medchemexpress LLC Plantainoside D | 147331-98-4 | 99.9% | 640.59 g/mol | C29H36O16 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN506310MG
Encompass_Preferred
Plantainoside D is a phenylethanoid glycoside research reagent reported to inhibit IKK-β and angiotensin-converting enzyme (ACE). It modulates voltage-dependent calcium channels and the PKC signaling cascade, reduces reactive oxygen species and NF-κB activation, and is used in studies of neuroprotection, anti-inflammation, antioxidant activity, and antihypertensive mechanisms.
  • Phenylethanoid glycoside with IKK-β and ACE inhibitory activity.
  • Modulates VDCCs and suppresses PKC signaling.
  • Reduces ROS generation and inhibits NF-κB activation.
  • High purity: 99.9%.
  • CAS number 147331-98-4.
  • Available as 10 MG solid and in DMSO solution formats for assays.
Catalog No. 50-002-89767