Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLAP Monoclonal Antibody (PLAP), Abbomax

Catalog No. 502204237
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4237 Supplier Abbomax Inc Supplier No. 604300
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Human kidney

PLAP Monoclonal antibody specifically detects PLAP in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen PLAP
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PLAP
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene ALPP
Gene Accession No. P05187
Gene Alias Alkaline phosphatase Regan isozyme; alkaline phosphatase, placental; alkaline phosphatase, placental type; alkaline phosphomonoesterase; ALP; ALPP; AP-TNAP; DOA1; FLJ11281; FLJ40094; FLJ61142; FLJ93059; glycerophosphatase; HOPS; MGC161443; MGC167935; P PLAP; PALP; PLA2P; PLAA; placental alkaline phosphatase 1; PLAP; PLAP-1; rCG23846-like; Regan isozyme; TNAP; TNSALP
Host Species Mouse
Immunogen A recombinant protein of human PLAP
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 250
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.