Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plas Labs™ Model 830 Compact Glove Box
GSA_VASDP

Catalog No. 11389121 Shop All Plas-Labs, Inc. Products
Change view
Click to view available options
Depth (English):
30 in.
31 in.
Dimensions (L x W x H):
30 x 49 x 31 in. (77 x 125 x 79 cm)
31 x 63 x 34 in. (78 x 160 x 86 cm)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English) Dimensions (L x W x H)
11-389-121 30 in. 30 x 49 x 31 in. (77 x 125 x 79 cm)
11-389-122 31 in. 31 x 63 x 34 in. (78 x 160 x 86 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-389-121 Supplier Plas Labs™ Supplier No. 830ABB
Add to Cart
Add to Cart
Request a Quote

Includes

White ambidextrous Hypalon™ gloves for superior chemical and U.V. resistance; Manual.

For safe containment of hazardous materials when being weighed

Work chamber accommodates analytical and toploading balances, bench scales and moisture analyzers in a clean, safe, portable and draft-free atmosphere.

  • Top section is removable for easy installation of equipment
  • Four purging gas valves (two on the transfer chamber and two on the main chamber)
  • White leveling tray for easy transferring of liquids
  • All clamps stainless steel and completely adjustable
  • Pressure relief valve minimizes glove fight back
  • HEPA filter on relief valve prevents escape of particulates and organic vapors
  • HEPA and organic vapor canister filters (replaceable)
  • Roomy transfer chamber is 12L x 11 in. I.D.

Certifications

Included hospital-grade multiple electrical outlet strip is UL listed, CSA approved.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Glove Box
Certifications/Compliance UL, CSA, CE
Depth (English) 30 in.
Depth (English) Interior 29 in.
Depth (Metric) 77 cm
Depth (Metric) Interior 74 cm
Dimensions (L x W x H) 30 x 49 x 31 in. (77 x 125 x 79 cm)
Height (English) 31 in.
Height (English) Interior 30 in.
Height (Metric) 79 cm
Length (English) 30 in.
Length (Metric) 77 cm
Voltage 120 V
Volume (English) 17.6 cu. ft.
Width (English) 49 in.
Width (English) Interior 35 in.
Width (Metric) 125 cm
Width (Metric) Interior 89 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.