Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Plasma Fluorescent Exosome Standard, Liquid, 100 μL

Catalog No. 502654714
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-4714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-4714 Supplier Abcam Supplier No. AB288123100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standardized plasma exosomes for EV tracking in fluorescence microscopy experiments.

  • Applications: Fluorescence Microscopy, Flow Cyt, EM
  • Form: Liquid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Exosomes
Shipping Condition Blue Ice
Form Liquid
Alternate Part Number M1073
Content And Storage Store at -20°C.
Detection Method Fluorescence
Sample Type Plasma
Quantity 100 μL
For Use With (Application) Fluorescence microscopy, flow cytometry, electron microscopy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.