Learn More
Creative Biomart Plasma protein fraction human (PPFh)
Supplier:  Creative Biomart THP0088
Encompass_Preferred
The product is defined as a sterile solution of proteins composed mainly of albumin and globulin derived from human plasma. This biologic should come from recovered plasma from whole blood or prepared source plasma. the final product should not include any type of additives. The composition of PPFh should be 5% of protein from which 83% should consist of albumin and no more of 17% should be globulin. It is also stated that no more than 1% of the total protein should be gamma globulin.PPFh is a sterile, frozen solution of solvent/detergent treated human plasma. The proteins in PPFh are stabilized with sodium caprylate and acetyltryptophan and it contains some electrolytes such as sodium, potassium and chloride.
Catalog No. 50-208-4139
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.