Learn More
Crystal Chem Inc Plasma Renin Activity (PRA) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80970
Encompass
The Plasma Renin Activity (PRA) ELISA is a highly sensitive assay that is used to quantify levels of human PRA in plasma.Highlights: Kit uses sample size of only 50 µL. High sensitivity of 0.15 ng/mL of Ang-I. Wide dynamic range of 0.20 - 60 ng/mL of Ang-I.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7873
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.