Learn More
Zymo Research Corporation Sample ZymoPURE II™ Plasmid Midiprep (2 preps)
Supplier: Zymo Research Corporation D4200S
Free Sample ZymoPURE™ technology uses a modified alkaline lysis method and features novel binding chemistry, which enables the highest yields and concentration of plasmid DNA (up to 3 g/l) directly from a spin-column. Coupling ZymoPURE with the innovative EndoZero™ II Spin-Columns, to eliminate endotoxins, achieves EndoZeroplasmid DNA ( 0.025 EU/g of plasmid DNA), making it suitable for transfection, restriction endonuclease digestion, in vivo studies, bacterial transformation, PCR amplification, DNA sequencing, and other sensitive downstream applications.As an added convenience, the ZymoPURE™ II Plasmid Midiprep Kit contains colored buffers that permit error-free visualization and identification of complete bacterial cell lysis and neutralization. Syringe filters are included for rapid clearing of the lysate and the unique spin-column design allows the binding step to be performed using a vacuum or table top centrifuge.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.