Learn More
Invitrogen™ Plasminogen Monoclonal Antibody (9130)

Description
MA1-43033 detects Plasminogen in mouse samples. MA1-43033 has successfully been used in Western blot and ELISA applications. MA1-43033 was derived from mouse plasminogen.
Specifications
Specifications
| Antigen | Plasminogen |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 9130 |
| Concentration | 6.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | PLG |
| Gene Accession No. | P20918 |
| Gene Alias | Ab1-346; Activation peptide; AI649309; angiostatin; DKFZp779M0222; Pg; plasmin; plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; plasminogen; PLG; RP1-81D8.1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.