Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plasminogen, MP Biomedicals™

Catalog No. MP219134250 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 U
250 U
50 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219134250 50 U
MP219134280 100 U
MP219134283 250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219134250 Supplier MP Biomedicals, Inc Supplier No. 0219134250
Add to Cart
Add to Cart

Found in human plasma (∼150mg/ml) and extracellular fluid. Plasminogen, MP Biomedicals is a single-chain plasma glycoprotein. Some activators (e.g. tissue plasminogen activator) convert plasminogen to its active form, plasmin. Plasminogen has been use

Two forms of plasminogen exist, namely the original form Glu-Plg and Lys-Plg. Activators such as tPA and scuPA are able to transform plasminogen into the two-chain plasmin, which expresses its enzymatic activity. Free plasmin in plasma will be inactivated immediately by its specific inhibitor, alpha-2-antiplasmin. Normally, plasmin only acts in the fibrin clot to degrade it.

Plasminogen is the inactive precursor of the protease plasmin. It is activated by the action of either tissue plasminogen activator (tPA), which primarily activates the fibrinolytic (thrombolytic) activity of plasmin, or urokinase plasminogen activator (uPA), which is associated with extracellular matrix remodeling and cell migration.

Plasmin cleaves fibrin/fibrinogen and blood coagulation factors V/Va and VIII/VIIIa. It activates matrix metalloproteinases by cleaving the inactive proenzymes. It is also involved in the activation of some growth factors, such as vascular endothelial growth factor (VEGF) and transforming growth factor β (TGF-β).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-91-6
Synonym Profibrinolysin

Specifications

Quantity 50 U
Formula Weight 90,000-94,000
Percent Purity ≥95%
Physical Form Solid
Recommended Storage −20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.