Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plasminogen Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534677 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534677 Supplier Invitrogen™ Supplier No. PA534677
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, Raji, K562, BCL-1, PC-12, Rat2, human plasma, mouse plasma, rat plasma. Predicted reactivity: Rat (80%), Dog (81%), Pig (80%), Rhesus Monkey (93%), Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plasminogen is a plasma protein synthesized mostly in the liver. It consists of a single polypeptide chain and has a molecular weight of 88.5 kDa. The plasma concentration is usually in the range of 70 - 200 mg/L. Plasminogen is activated by tissue-plasminogen activator, urokinase and streptokinase, to form plasmin. Activation results from the cleavage and release of the preactivation peptide. Plasmin consists of a heavy (A-) chain which contains 5 kringles and a light (B-) chain with the active site. Plasmin is a trypsin-like protease whose natural substrate is fibrin, with binding sites for fibrin residing on the kringles. Plasmin is essential for fibrinolysis and decreased fibrinolytic potential due to congenital defects in plasmin can lead to recurring thrombosis, thrombophlebitis and pulmonary embolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plasminogen
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7
Gene PLG
Gene Accession No. P00747, P20918, Q01177
Gene Alias Ab1-346; Activation peptide; AI649309; angiostatin; DKFZp779M0222; Pg; plasmin; plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; plasminogen; PLG; RP1-81D8.1
Gene Symbols PLG
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Plasminogen. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18815, 5340, 85253
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.