Learn More
Market Lab Plastic Box with Foam Tube Rack, 14" x 8" x 2.5
Supplier:  Market Lab 5875
Encompass_Preferred
The Plastic Box with Foam Tube Rack from MarketLab is a plastic storage box designed to hold up to 25 tubes. It measures 14" x 8" x 2.5" and is made from durable polypropylene plastic. The box features a foam insert with 25 holes to keep tubes neatly organized and protected. This storage box allows you to conveniently transport and store tubes in the laboratory, clinic, hospital or office. The secure lid keeps contents protected.
Catalog No. NC2832431
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.