Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Plastic Carboy with Gasket Cap
SCI_ED

Catalog No. S27493 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S27493 10 L
S27494 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27493 Supplier Eisco™ Supplier No. CH0065A
Add to Cart
Add to Cart

Made of lab quality polypropylene plastic, designed for holding large amounts of liquid regularly used in the laboratory

  • Premium polypropylene plastic carboys
  • Ideal for any laboratory
  • Gasket Lids seal the container
  • Significantly lighter than glass, highly portable, and durable
  • Handles provide easy maneuverability and placement
  • Autoclavable

Specifications

Autoclavable Autoclavable
Capacity (Metric) 10 L
Closure Material Plastic
Closure Type Screw Cap with Gasket
Color White
Disposable Reusable
For Use With (Application) Holding Liquid
Graduations No
Material Polypropylene
Mouth Style Narrow
Neck Style Short
Product Type Carboy
Shape Cylindrical
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.