Learn More
American Educational Products Plastic Column Kit
SCI_ED
Catalog No. S53037 Shop All American Educational Products LLC Products
Change view
Click to view available options
Product Type:
7 mm Beads
Catalog No. Product Type
S53037 7 mm Beads
1 options
Catalog No. S53037 Supplier American Educational Products Supplier No. 3276
Includes
tube, drainage fixture, screen, and teacher's guide.
Learn how the movement of water is affected by substrate!
How do porosity and permeability affect the distribution of aquifers? Help students study these important concepts and integrate math measurement and percentage skills. Bead set is optional. Grades 6-12.
Specifications
Grade 6 to 12
Product Type 7 mm Beads