Learn More
Fisherbrand™ Plastic Disposable Generator Probe
12 mm Fisherbrand Plastic Disposable Generator Probe significantly reduces the complexity of disassembly and maintenance compared to traditional stainless-steel probes. Made from durable plastic, these tips are cost-effective for single use, minimizing the risk of cross-contamination.
 View more versions of this product
Catalog No. 15-341-135
Only null left
Add to Cart
Description
Description
  • Suitable for homogenization where sample size or sample hardness dictates that processing with the 7 mm tip is prohibitive.
  • Significantly reduces cross-contamination and is virtually maintenance-free
  • Suitable for PCR, RNA, and DNA extraction
  • Compatible with Fisherbrand™ 150 Handheld Homogenizer, Fisherbrand™ 850 Homogenizer, Omni Tissue Homogenizer, Omni THQ, and Omni GLH 850 homogenizer
  • Provides a cost-effective solution for researchers homogenizing samples where contaminants can affect downstream assays
Specifications
Specifications
Probe
Autoclavable
Blue
12 mm
Polypropylene
50/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.