Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keck Plastic Joint Clips

Catalog No. 05880C Shop All Oakton Products
Change view
Click to view available options
For Use With (Equipment):
Joint size 14
Joint size 19
Joint size 24
Color:
Blue
Green
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Color
05-880C Joint size 14 Yellow
05-880D Joint size 19 Blue
05-880E Joint size 24 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-880C Supplier Keck Supplier No. 0684005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Made of plastic for use on standard taper glass joints

Made of plastic for use on standard taper glass joints

  • Color coded for easy size identification
  • Cost less than metal clips
  • Do not corrode
  • Will not scratch glass

Specifications

Type Taper Clamp
Material POM
For Use With (Equipment) Joint size 14
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.