Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Plastic Test Tube Rack, 3-Tier
SDPSCI_ED

Catalog No. S15729 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Positions:
12
18
36
40
62
For Use With (Equipment):
13 mm Tubes
16 mm Tubes
18 mm Tubes
20 mm Tubes
25 mm Tubes
32 mm Tubes
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions For Use With (Equipment)
S15729 40 20 mm Tubes
S15979 18 13 mm Tubes
S15981 18 25 mm Tubes
S16081 62 16 mm Tubes
S15958 12 25 mm Tubes
S16006 36 25 mm Tubes
S15731 12 32 mm Tubes
S15851 12 18 mm Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S15729 Supplier United Scientific™ Supplier No. 77719
Only null left
Add to Cart
Add to Cart

Can be disassembled to store flat

  • Polypropylene racks with a 3-tier design provide a clear view of the tube contents and convenient insertion and removal of tubes.
  • Available in multiple diameters and configurations.

Specifications

Autoclavable Autoclavable
Color White
Holds Test Tubes
Material Polypropylene
Positions 40
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) 20 mm Tubes
Includes 2 Racks
Product Type Test Tube Rack (3-tier)
Shelves 3 Tiers
Array 4 x 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.