Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Plastic Test Tube Rack
SDPSCI_ED

Catalog No. S23992 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Positions:
8 and 36
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
S23992 8 and 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23992 Supplier United Scientific Supplies Supplier No. 76001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choose a versatile test tube rack. The Plastic Test Tube Rack features four sides, for 12 mm to 25 mm diameter test tubes.

  • All four sides can be used interchangeably for 12 mm through 25 mm diameter tubes
  • Rack features a total of 88 spaces on its four sides, 8 for 50 mL, 20 for 20 mL, 24 for 15 mL, and 40 for 12 mL tubes

Specifications

Autoclavable Autoclavable
Color Blue
Holds Tubes (12 and 25 mm)
Material Polypropylene
Positions 8 and 36
Shape Cubical
Stackable No
Sterility Non-sterile
For Use With (Equipment) Can be used with 12 mm through 25 mm diameter tubes
Diameter (Metric) Opening 12 mm and 25 mm
Product Type Test Tube Rack
Shelves None
Array 88 spaces on its four sides, 8 for 50 ml, 20 for 20 mL, 24 for 15 mL, and 40 for 12 mL tubes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.