Learn More
United Scientific™ Plastic Test Tube Rack, In-Line
SDPSCI_ED
Catalog No. S01306 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Positions:
6
Catalog No. Positions
S01306 6
1 options
Catalog No. S01306 Supplier United Scientific™ Supplier No. TTHP02
Add to Cart
Add to Cart
In-Line Plastic Test Tube Rack is fully assembled and ready to use right out of the box.
  • This polyethylene rack will hold six test tubes, and has six drying pins.
  • Sides are fully open for clear viewing.
  • Rack has six 25 mm holes.
Specifications
Autoclavable Not Autoclavable
Color White
Holds PCR Tubes
Material Polyethylene
Positions 6
Shape Horizontal
Stackable No
Sterility Non-sterile
Diameter (Metric) Opening 25 mm
Product Type Test Tube Holder
Shelves None
Array 6