Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Plastic Test Tube Rack, Wet/Dry
SDPSCI_ED

Catalog No. S01226 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Positions:
21
24
40
60
90
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
S01226 60
S01225 90
S01227 40
S01228 24
S01229 21
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S01226 Supplier United Scientific™ Supplier No. 77902
Add to Cart
Add to Cart
This item is not returnable. View return policy

Test tube racks that can be used in water.

  • Economical yet very durable racks that are submersible, autoclavable, and easily stack when empty
  • Made from a special blend of polypropylene (PP) that makes them sink in a water bath and maintain their stability, even in agitated water
  • The top tier features an alphanumeric grid reference
  • Available in white, but can be special ordered in various colors
  • Shipping weight (for pack of 6) 2.0 lb.
  • Package dimensions: 15 x 10 x 2 in.

Specifications

Autoclavable Autoclavable
Color White/White
Holds Tubes (16 mm)
Material Polypropylene
Positions 60
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Test Tubes
Includes 6 Racks
Product Type Test Tube Rack
Shelves None
Array 5 x 12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.