Learn More
Youngstown Barrel & Drum Plastic Tight Head Drums

Description
- Made of chemical-resistant high-density polyethylene
- Tight-head drums for transport of hazardous liquids
Specifications
Specifications
| Certifications/Compliance | 1H1/Y1.9/150 |
| Product Type | Plastic Tight Head Drums |
| Capacity (English) | 30 gal. |
| Capacity (Metric) | 113.56 L |
| Color | Blue |
| For Use With (Application) | For Transport of Hazardous Solids or Liquids |
| Material | Plastic |
| Freight Class | 50 |
| Head Style | Tight |
| Height (English) | 29.6 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.