Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic Tight Head Drums
SDP

Catalog No. 19058096
Change view
Click to view available options
Color:
Blue
Natural
Certifications/Compliance:
1H1/Y1.9/150
UN 1H1/Y1.8/100
UN 1H1/Y1.9/100
UN 1H1/Y1.9/150
Diameter (English) Outer:
14.2 in.
19.25 in.
19.5 in.
23.3 in.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Certifications/Compliance Diameter (English) Outer
19-058-096 Blue UN 1H1/Y1.9/100 23.3 in.
19-058-099 Blue UN 1H1/Y1.8/100 14.2 in.
19-058-098 Blue UN 1H1/Y1.8/100 19.25 in.
19-058-095 Natural UN 1H1/Y1.9/100 23.3 in.
19-058-097 Natural UN 1H1/Y1.9/150 19.25 in.
19-058-100 Natural UN 1H1/Y1.8/100 14.2 in.
18-937A Blue 1H1/Y1.9/150 19.5 in.
18-936A Blue UN 1H1/Y1.9/150 23.3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-058-096 Supplier Youngstown Barrel & Drum Supplier No. 1295M
Add to Cart
Add to Cart

High-density polyethylene containers for transport of hazardous solids or liquids

  • Made of chemical-resistant high-density polyethylene
  • Tight-head drums for transport of hazardous liquids

Specifications

Certifications/Compliance UN 1H1/Y1.9/100
Product Type Plastic Tight Head Drums
Capacity (English) 55 gal.
Capacity (Metric) 208.19 L
Color Blue
For Use With (Application) For Transport of Hazardous Solids or Liquids
Material Plastic
Freight Class 50
Head Style Tight
Height (English) 34.8 in.
Height (Metric) 88.39 cm
Diameter (English) Outer 23.3 in.
Diameter (Metric) Outer 59.18 mm
Weight (English) 21.5 lb.
Weight (Metric) 9.75 Kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.