Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Plastic Tweezers

Catalog No. 19090651 Shop All Medique Products
Change view
Click to view available options
Quantity:
100/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-651 100/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-090-651 Supplier Medique Supplier No. 77233
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Emergency preparedness instrument for use in any medical situation.

  • Blue color
  • Disposable

Specifications

For Use With (Application) Emergency Medical Situation
Product Type Disposable Tweezers
Length (English) 4.5 in.
Color Blue
Material Plastic
Quantity 100/Bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.