Learn More
Lakeside Manufacturing Inc Plastic Utility Cart 500Lb Chr
Supplier:  Lakeside Manufacturing Inc 2512
Encompass_Preferred
Plastic Utility Carts by Lakeside Manufacturing with an adjustable middle shelf offer generous flexibility and convertibility when loading cart. The Lakeside 2509 Plastic Utility Cart with (3) 16-1/4" x 28-3/4" shelves has a weight capacity of 300-Lb. This Lakeside cart's adjustable middle shelf offers generous convertibility when loading cart. This cart's sturdy polyethylene construction is stain, odor, and impact-resistant. This cart's integrated push handle allows for comfortably guiding cart with ease. This utility cart's no-mark, all-swivel casters, 2 with brakes, provide ultimate maneuverability. The medical utility cart's zinc-plated legs are sleek and durable. Its seamless shelves with contoured corners are contemporary and easy to clean and sanitize. Assembles easily without hardware. Comes standard in white. Modifiable to meet your needs. Superior lead times. Made in China.
Catalog No. 50-288-4407
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.