Learn More
Lakeside Manufacturing Inc Plastic Utility Cart 500Lb Gry
Supplier: Lakeside Manufacturing Inc 2510
Plastic Utility Carts by Lakeside Manufacturing with deep well shelves hold medical supplies and equipment securely during transport. The Lakeside 2507 Plastic Utility Cart with (3) 16-1/4" x 28-3/4" deep well shelves has a weight capacity of 300-Lb. This Lakeside cart's deep well shelves hold and transport items securely. This cart's sturdy polyethylene construction is stain, odor, and impact-resistant. This cart's integrated push handle allows for comfortably guiding cart with ease. This utility cart's no-mark, all-swivel casters, 2 with brakes, provide ultimate maneuverability. The medical utility cart's black anodized aluminum legs are sleek and durable. Its seamless shelves with contoured corners are contemporary and easy to clean and sanitize. Assembles easily without hardware. Comes standard in charcoal. Modifiable to meet your needs. Superior lead times. Made in China.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.