Learn More
Lakeside Manufacturing Inc Plastic Utility Cart, Charcoal
Supplier:  Lakeside Manufacturing Inc 2503
Encompass_Preferred
Plastic Utility Carts by Lakeside Manufacturing are a contemporary and economical piece of equipment that makes for an easy transportation of light-duty medical equipment and supplies. The Lakeside 2500 Plastic Utility Cart with (3) 16" x 25-1/2" shelves has a weight capacity of 300-Lb. This Lakeside cart's stain and odor resistant material maintains an attractive appearance, while its sturdy polyethylene construction is impact-resistant. This cart's integrated dual push handles allow for comfortably guiding cart with ease. This utility cart's no-mark, all-swivel casters, 2 with brakes, provide ultimate maneuverability. The medical utility cart's epoxy coated steel legs are sleek and durable. Easy to clean and sanitize. Assembles easily without hardware. Comes standard in black. Modifiable to meet your needs. Superior lead times. Made in China.
Catalog No. 50-288-4475
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.