Learn More
Plasticoid Bailey Rubber Crucible Holder
SDP
Catalog No. 08265 Shop All Plasticoid Products
Change view
Click to view available options
Material:
Rubber
Catalog No. Material
08-265 Rubber
1 options
Catalog No. 08-265 Supplier Plasticoid Supplier No. VCHBAL
Add to Cart
Add to Cart
Bailey Rubber Crucible Holder. Fits top of 50mm funnels. For 08-195D and -E Gooch Crucibles (below, left). Wide part of holder rests on top of funnel. Projection rests on inside and supports holder with suction. Rubber part only.
Specifications
Material Rubber
Product Type Crucible Holder