Learn More
Description
Contents:
- 1 Five-piece Beaker Set (50, 100, 250, 500 & 1000)
- 1 Four-piece Graduated Cylinder Set (10, 25, 50 & 100 mL)
- 1 Wash Bottle (250 mL)
- 1 Wash Bottle (500 mL)
- 1 Narrow-mouth Reagent Bottle, PP (250 mL)
- 1 Narrow-mouth Reagent Bottle, PP (500 mL)
- 1 Wide-mouth Reagent Bottle, PP (250 mL)
- 1 Wide-mouth Reagent Bottle, PP (500 mL)
- 1 Measuring Jug (500 mL)
- 1 Measuring Jug (1000 mL)
Specifications
Specifications
| Capacity (Metric) | 0.05 to 1 L |
| Material | Polypropylene |
| For Use With (Application) | General Lab Use |
| Type | Plasticware Starter Kit |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
| Scale | Molded Graduations |
| Style | Assorted |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

