Learn More
Thomas Scientific Krystal 2000 96 Well Microplate, Black, Bottom Flat, 350æl Well Capacity, Qty: 100
Supplier: Thomas Scientific 1158A52
"No Cross Talk" Opaque Clear Bottom 96 Well Plate These plates should be used when light cross talk is important to avoid. The patented manufacturing process creates 96 individual clear wells in an opaque plate matrix, completely eliminating light cross talk from well to well. Applications include fluorescence, luminescence and scintillation. Standard, high bind treated for protein, or tissue culture treated versions available. Designed to SBS specifications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.