Learn More
Thomas Scientific 96 Well Solid Microplates, Polystyrene, clear, Round well bottom, 325æl Capacity, Inner Pack: 10, 100/PK
SDP
Supplier:  Thomas Scientific 1158A44
Encompass
350ul Well Volume These 96 well solid microplates are available in black, white, and clear polystyrene or natural polypropylene. The colored plates are ideal for fluorescence, luminescence and scintillation while the clear plates are useful for ELISA assays. Designed to SBS specifications.
Catalog No. 50-146-0872
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.