Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Plated Media Trypticase™ Soy Agar

Catalog No. 22350895 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
32 mL
100 mL
Color:
Light Beige
Light Yellow
Media Type:
Trypticase™ Soy Agar
Packaging Type:
Bottle
Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color Media Type Packaging Type
22-350-895 32 mL Light Yellow Trypticase™ Soy Agar Plate
B4392385 100 mL Light Beige Trypticase™ Soy Agar Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-350-895 Supplier BD Supplier No. 215297
Add to Cart
Add to Cart
This item is not returnable. View return policy

For isolation and cultivation of non-fastidious and fastidious microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Yellow
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar
Packaging Type Plate
Quantity 32 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.