Learn More
Research Products International Corp Deep Well Plates, S, 384 Wells, Square V, 240ul, 5/pk
SDP
Supplier:  Research Products International Corp 162526S
Encompass_Preferred
Deep Well Plates are ideal for sample collection, preparation and long-term STOR. Raised rim on each well facilitates secure sealing with film or sealing mat. Plates allow optimum sample mixing and recovery through highly polished walls and offer reliable automation performance. Wells feature alphanumeric indexing molded in on the A-H and 1-12 axis with maximum centrifugation to 4,000 RCF.Manufactured from virgin polypropylene (autoclavable) and are ultra-clear for easy visualization of samples. Plates are compatible with most liquid handling robots and SBS footprint meets ANSI/SLAS 1-2004. Stackable and flatness tested for reliable automation performance.Capacity (ul): 240Wells: 384 Square, V-BottomIndexing: Clear alphanumericRNase, DNase, DNA, PCR Inhibitor freeNon-SAutoclavableStackableUltra-clearHTS friendlyFootprint meets ANSI/SLAS 1-2004Shipping Weight: 2 lbsProduct usually ships within 24 hours.Research or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-0159
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.