Learn More
Eppendorf™ Platform for Innova™ 2300/43/43R, Excella™ E10/E25/E25R
Shop All Eppendorf North America ProductsDescription
- Platform for Innova™ 2300/43/43R, Excella™ E10/E25/E25R
- 76 x 46 cm (30 x 18 in.)
- Aluminum
- Erlenmeyer flask dedicated platform
Specifications
Specifications
| Holds | 15 x 1 L Erlenmeyer Flasks |
| Product Type | Erlenmeyer Flask Platform |
| Material | Aluminum |
| Dimensions (L x W) | 76 x 46 cm (30 x 18 in.) |
| For Use With (Equipment) | Innova™ 2300/43/43 R, Excella™ E10/E25/E25R |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.