Learn More
Grainger K4796 Silicone Round Cord 1/16 D 50 L 70A Blue
Supplier:  Grainger 16Z139
Encompass_Preferred
The Silicone Round Cord Metal is a silicone rubber cord with a metal core. It is made of high-quality silicone rubber that is flexible and durable. The cord has a round cross section and comes in multiple diameters from 1/8 inch to 3/4 inch. It can withstand temperatures from -85°F to 500°F. The metal core provides strength while allowing the cord to remain flexible. Common applications include gasketing, sealing, vibration dampening, noise reduction, and impact absorption. This product can be cut to length with scissors or a knife. It is RoHS compliant and made in the USA.
Catalog No. 50-276-8018
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.