Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ Hot Start PCR Master Mixes (2X)

Catalog No. 13001013
Encompass_Preferred
Change view
Click to view available options
Color:
Colorless
Green
No. of Reactions:
1000 Reactions
200 Reactions
50 Reactions
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Reactions
13-001-013 Green 200 Reactions
13-000-012 Colorless 50 Reactions
13-000-013 Colorless 200 Reactions
13-000-014 Colorless 1000 Reactions
13-001-012 Green 50 Reactions
13-001-014 Green 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13-001-013 Supplier Invitrogen™ Supplier No. 13001013
Only null left
Add to Cart
Edge
Add to Cart

Invitrogen Platinum Hot Start PCR Master Mix offers Platinum Taq DNA polymerase in an optimized PCR buffer with magnesium and dNTPs for convenient PCR setup.

Invitrogen Platinum Hot Start PCR Master Mix, available in colorless and green formats, offers Platinum Taq DNA polymerase in an optimized PCR buffer with magnesium and dNTPs for convenient PCR setup. It retains all the features of Platinum Taq DNA Polymerase: specificity, robustness, and reliability.

Platinum Green Hot Start PCR Master Mix additionally contains a density reagent and two tracking dyes for direct loading of PCR products on gels.

Platinum Hot Start PCR Master Mix provides a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs, thus reducing the number of pipetting steps during PCR reaction setup. It is supplied with a separate vial of Platinum GC Enhancer, formulated for specific amplification and improved yields of GC-rich targets.

Platinum Taq DNA Polymerase is a recombinant Taq DNA polymerase complexed with proprietary antibodies that inhibit enzyme activity at room temperature, reducing nonspecific target amplification at lower temperatures. The antibodies dissociate during the initial PCR denaturation step, and DNA polymerase regains its full activity. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template-dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity.

Features

  • High specificity and increased yields with antibody-mediated hot-start PCR
  • Convenient room temperature reaction setup
  • Versatile formulation for broad range of amplicons
  • Direct gel loading with green PCR master mix

Applications

  • Hot Start PCR
  • Genotyping
  • Gene expression profiling
  • Next-generation sequencing

Notes

  • Platinum II Taq Hot-start DNA Polymerase is recommended superior PCR performance.
  • For applications that require PCR product analysis by absorbance or fluorescence excitation, colorless format is recommended.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • Platinum Green Hot Start PCR 2X Master Mix, 4 x 1.25 mL
• Platinum GC Enhancer, 2 x 1.25 mL
• Nuclease-free water, 4 x 1.25 mL

Store at -20°C in a non-frost-free freezer.
Format Tube
GC-Rich PCR Performance Medium
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Standard
Label or Dye Green
Product Type Hot Start PCR Master Mix
Quantity 200 reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Size (Final Product) 5 kb or less
Starting Material DNA
Color Green
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.