Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ Tfi Exo(-) DNA Polymerase

Catalog No. 60684050
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-684-050 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-684-050 Supplier Invitrogen™ Supplier No. 60684050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Platinum™ Tfi Exo(-) DNA Polymerase

Platinum™ Tfi Exo(-) DNA Polymerase is recombinant Tfi Exo(-) DNA Polymerase complexed with a proprietary antibody mix that inhibits polymerase activity at ambient temperature, allowing room-temperature reaction setup. Activity is restored after the initial denaturation step in PCR cycling at 94°C, providing an automatic 'hot start' for increased specificity, sensitivity, and yield. Tfi Exo(-) DNA Polymerase is purified from E. coli. expressing cloned mutants of the Thermus filiformis DNA polymerase gene. This enzyme has 5'→3' polymerase activity, but lacks both 5'→3' and 3'→5' exonuclease activity for improved yield and robustness. Tfi Exo(-) DNA polymerase is heat-stable and will synthesize DNA at elevated temperatures from single-stranded templates in the presence of a primer.

Applications
Platinum™ Tfi Exo(-) DNA Polymerase can be used in protocols that currently use Platinum™ Taq DNA Polymerase without modification. PCR performance is comparable to that of Platinum™ Taq in yield, specificity, fidelity, and robustness. Like Platinum™ Taq, Platinum™ Tfi Exo(-) DNA Polymerase has a nontemplate-dependent terminal transferase activity that adds a single deoxyadenosine (A) to the 3' ends of PCR products.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage Platinum™ Tfi DNA Polymerase (500 units total, at 5U/μL) is supplied with 5X Platinum™ Tfi Reaction Buffer (4 × 1.3 mL) and 50 mM Magnesium Chloride (1 mL). Store at -20°C (non-frost-free).
GC-Rich PCR Performance Low
Polymerase Tfi DNA Polymerase
Reaction Speed Standard
Product Line Platinum
Product Type DNA Polymerase
Quantity 500 units
Shipping Condition Dry Ice
Fidelity (vs. Taq) 1 X
Hot Start Built-In Hot Start
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 1 kb or less
Volume 5.2 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.