Learn More
Description
Includes
40 μL Platinum II Taq HS DNA Polymerase, 1.25 mL 5X Platinum II PCR Buffer, 1.25 mL Platinum GC Enhancer
Invitrogen Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology.
Features of Platinum II Taq Hot-Start DNA Polymerase include:
- Innovative buffer—enables universal annealing temperature by isostabilizing primer-template duplex structures
- Engineered Taq DNA polymerase—confers fast cycling and resistance to common inhibitors
- Platinum hot-start technology—enables superior specificity, sensitivity, and yields; allows for room temperature reaction setup
Platinum II Taq Hot-Start DNA Polymerase is an engineered Taq DNA polymerase that shows increased resistance to reaction inhibitors originating from sample material or DNA purification steps. The polymerase has a higher DNA synthesis rate and delivers PCR results more than two times faster than other Taq DNA polymerases. Proprietary Platinum Taq antibodies block polymerase activity at ambient temperatures and dissociate after the initial denaturation step at 94°C. This automatic 'hot start' provides increased sensitivity, specificity, and yield, while allowing reaction assembly at room temperature.
Due to the unique composition of the Platinum II PCR buffer, the annealing temperature is 60°C for most primer pairs designed following the general design rules. Isostabilizing molecules in the buffer increase primer–template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize annealing temperature for each primer pair. With Platinum II Taq Hot-Start DNA Polymerase, different PCR assays can be cycled together using the same protocol with universal primer annealing temperature and the extension step selected for the longest fragment to be amplified.
Platinum II Taq Hot-Start DNA Polymerase is provided with the optional Platinum GC Enhancer for specific amplification and improved yields of GC-rich targets.
Use Platinum II Taq Hot-Start DNA Polymerase for the amplification of DNA from complex genomic, viral, and plasmid templates, as well as in RT-PCR, in applications like genotyping, high-throughput PCR, or with samples of suboptimal purity.
For increased convenience, we offer Platinum II Hot-Start PCR Master Mix (2X) (Cat. No. 14000012), where Platinum II Taq Hot-Start DNA Polymerase is provided in a ready-to-use mixture with Platinum II PCR buffer and dNTPs, thus reducing the number of pipetting steps during PCR reaction setup. Platinum II Hot-Start Green PCR Master Mix (2X) is also available, which additionally contains a density reagent and two tracking dyes for direct loading of PCR products on gels, further streamlining the PCR workflow from setup to final analysis of the result.
Applications:
- Genotyping and allele screening
- Amplification from low-input or low-quality DNA
- GC-rich and AT-rich PCR
- Sanger sequencing template generation
- Broad-range microbial/16S amplification
- Fast and high-throughput PCR
- Multiplex target amplification
- Amplification of challenging genomic targets
- Colony PCR and clone screening
- qPCR-based pathogen detection and quantification
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | • Platinum II Taq HS DNA Polymerase, 40 μL • 5X Platinum II PCR Buffer, 1.25 mL • Platinum GC Enhancer, 1.25 mL Store at-20°C in a non-frost-free freezer. |
| Format | Tube |
| GC-Rich PCR Performance | High |
| Polymerase | Platinum II Taq Hot-Start DNA Polymerase |
| Reaction Speed | Fast or Standard |
| Product Type | Hot Start DNA Polymerase |
| Quantity | 100 reactions |
| Shipping Condition | Room Temp or Wet Ice |
| For Use With (Application) | Hot-start PCR |
| Show More |
Frequently Asked Questions (FAQs)
Platinum II Taq Hot-Start DNA Polymerase products can be stored at 4 degrees C for up to 3 months. For longer storage, we recommend storing all components at -20 degrees C.
No. The tracing dyes (a blue and a yellow dye) in Platinum II Green PCR Buffer do not interfere with fragment separation on E-Gel agarose gels.
No. The tracing dyes (a blue and a yellow dye) in Platinum II Green PCR Buffer do not interfere with PCR performance and do not change any enzyme features.
Yes, Platinum II Taq Hot-Start DNA Polymerase can be used in qPCR master mixes for target detection and quantification in a real-time PCR instrument using probes or SYBR Green dye.
Yes. Due to stable antibody-mediated hot-start technology, Platinum II Taq Hot-Start DNA Polymerase is highly stable. The premixed reactions for PCR can be incubated at room temperature for up to 24 hr before loading in the thermal cycler, without any loss of amplification specificity.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.