Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)

Catalog No. AA41802FI
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41802FI 50 x 50 mm
AA41802FF 25 x 25 mm
AA41802GH 100 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41802FI Supplier Thermo Scientific Chemicals Supplier No. 041802FI

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

Used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings. | Pt:Ir; 90:10 wt%

Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis) is used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Iridium foil
Quantity 50 x 50 mm
Molecular Formula Pt:Ir; 90:10 wt%
MDL Number MFCD02091683
Solubility Information Insoluble in water.
Percent Purity 99.9%
Packaging Tube

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.