Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum Iridium gauze, 150 mesh woven from 0.043mm (0.0017in) dia wire

Catalog No. AA40934FF
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40934FF 25 x 25 mm
AA40934FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40934FF Supplier Thermo Scientific Chemicals Supplier No. 040934FF

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

Filtration, electrodes

Filtration, electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Iridium gauze
Name Note Woven from 0.043mm (0.0017 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
Quantity 25 x 25 mm
Mesh Size 150 Mesh
Odor Odorless
Form Wire Cloth
MDL Number MFCD02091683

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.