Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(IV) bromide, 99.99% (metals basis), Pt 37.1% min, Thermo Scientific Chemicals

Catalog No. p-7048947
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1217103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1217103 Supplier Thermo Scientific Chemicals Supplier No. 01217103

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

Platinum(IV) bromide can be used as an analytical reagent to test for the presence of cesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68938-92-1
Molecular Formula Br4Pt
Molecular Weight (g/mol) 514.70
MDL Number MFCD00016289
InChI Key DYFRBYUUSCNRBP-UHFFFAOYSA-J
Synonym platinum tetrabromide, platinum iv bromide, platinum 4+ tetrabromide
IUPAC Name tetrabromoplatinumtetrakis(ylium)
SMILES Br[Pt+4](Br)(Br)Br

Specifications

Melting Point 180°C (decomposition)
Color Brown to Black
Physical Form Crystalline
Quantity 1 g
Linear Formula PtBr4
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 514.73
Percent Purity 99.99%
Concentration or Composition (by Analyte or Components) Pt 37.1% min.
Assay (metals basis)
Chemical Name or Material Platinum(IV) bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 273-151-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.