Learn More
Sigma Organic Chemistry Platinum Nanospheres Citrate Buffer | 25ML | 007440-06-4
Supplier:  Sigma Organic Chemistry 91605625ML
Encompass_Preferred
Platinum Nanospheres Citrate Functionalized 5 nm Diameter 0.05 mg/ml In 2 mm Aqueous Sodium Citrate Buffer, 25ml
About This Item:
Unspsc Code: 12352200
Storage: Cooler/Refrigerated
Catalog No. 91-605-625ML
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.