Learn More
Invitrogen™ Platinum™ Taq DNA Polymerase
Description
Includes
Each box contains: Platinum Taq DNA Polymerase; 10X PCR Buffer (no magnesium chloride); 50mM magnesium chloride; KB Extender
Invitrogen Platinum Taq DNA Polymerase is a convenient and reliable hot-start thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase. The hot start property of the enzyme is conferred by thermolabile monoclonal antibodies that render Taq DNA polymerase inactive until the initial PCR denaturation step, thus preventing the extention of nonspecifically annealed primers and improving product yield.
The hot-start property allows for convenient reaction assembly at room temperature. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity. PCR products generated with Platinum Taq DNA Polymerase may be used in the same downstream applications without protocol modifications.
Features
- Convenient reaction assembly at room temperature
- 5' to 3' exonuclease activity
- Lacks 3' to 5' exonuclease activity
Applications
- Amplification of DNA from complex genomic, viral, and plasmid templates
- Hot-start PCR
- RT-PCR
Notes
- For superior PCR performance, the next-generation enzyme Platinum II Taq Hot-start DNA Polymerase is recommended.
- Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology.
Order Info
Shipping Condition: Approved for shipment on wet or dry ice
Specifications
Specifications
| Content And Storage | • Platinum Taq DNA Polymerase, 60 μL • 10X PCR Buffer (without magnesium), 2 x 1.25 mL • 50 mM MgCl2, 1 mL • KB Extender, 1.25 mL Store at -10°C to -30°C in a non-frost-free freezer. |
| Format | Tube |
| GC-Rich PCR Performance | Low |
| Polymerase | Platinum Taq DNA Polymerase |
| Reaction Speed | Standard |
| Exonuclease Activity | 5' - 3' |
| Product Type | DNA Polymerase |
| Quantity | 300 reactions |
| Shipping Condition | Wet or Dry Ice |
| For Use With (Application) | Hot-start PCR |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.