Learn More
Thomas Scientific Platinum Wire, 32 Gauge, Per Inch
SDP
Supplier:  Thomas Scientific 1233S71
Encompass
Platinum Wire can be used in several applications such as gasket or o-ring fabrication, thermocouples, electrical contacts, and heater coils. • Gage specified is B. and S.; weights in milligrams per inch are approximate. • Gage, Mg/inch: 24:108; 25:92; 26:71; 28:46 and 36:7.
Catalog No. NC0273494
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the Fisher Scientific channel.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.