Learn More
Platypus Technologies LLC Platypus Technologies Gold-Coated Microscope Slides, 1"x3" Aluminosilicate, 100 nm Gold Film, 5 Slides Per Pack
Supplier: Platypus Technologies LLC AU.1000.ALSI
Gold-coated microscope slides are made from aluminosilicate glass (1-inch x 3 inch x 0.7 mm) coated with a film of gold (100-nm thick) via e-beam evaporation. A 2.5 nm film of titanium is used as an adhesion layer between the gold film and the glass. Purity of gold is 99.999%. Package contains 5 gold-coated microscope slides. Gold-coated microscope slides are ideal as electrodes for potentiometry and biosensing, infrared (IR) spectroscopy, Raman spectroscopy or ellipsometry. Pristine gold can be readily functionalized with thiol-terminated self-assembled monolayers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.